Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026305.2 | NP_080581.1 | 4 | 360 | ||
GenBank mRNA | AK002223.1 | - | 4 | 382 | ||
AK002868.1 | - | 4 | 361 | |||
AK003277.1 | - | 4 | 352 | |||
AK008477.1 | - | 4 | 369 | |||
AK008534.2 | - | 4 | 363 | |||
AK012254.1 | - | 4 | 353 | |||
AK019181.1 | - | 4 | 388 | |||
AK019218.1 | - | 4 | 388 | |||
AK019358.1 | - | 4 | 353 | |||
AK152950.1 | - | 5 | 431 | |||
BC051927.1 | - | 3 | 346 | 5684198 | ||
BC056983.1 | - | 3 | 345 | 6823854 |