Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026310.3 | NP_080586.1 | 3 | 519 | ||
GenBank mRNA | AK003104.1 | - | 3 | 524 | ||
AK008680.1 | - | 3 | 520 | |||
AK010150.1 | - | 3 | 572 | |||
AK089039.1 | - | 2 | 228 | |||
BC011425.1 | - | 3 | 494 | 3603233 |