Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kirrel3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190911.1 | NP_001177840.1 | 2, 3 | 514 | ||
NM_001190912.1 | NP_001177841.1 | 1, 2 | 436 | |||
NM_001190913.1 | NP_001177842.1 | 2, 3 | 514 | |||
NM_001190914.1 | NP_001177843.1 | 3, 4 | 615 | |||
NM_026324.3 | NP_080600.1 | 2, 3 | 514 | |||
GenBank mRNA | AK034142.1 | - | 2, 3 | 451 | ||
AK038385.1 | - | 2, 3 | 265 | |||
AK045373.1 | - | 2, 3 | 419 | |||
AK079332.1 | - | 3, 4 | 552 | |||
AK122566.1 | - | 1, 2 | 28 | |||
AY169782.1 | - | 2, 3 | 514 | |||
BC063072.1 | - | 1, 2 | 231 | 5695003 |