Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162368.1 | NP_001155840.1 | 6 | 726 | ||
NM_001162369.1 | NP_001155841.1 | 6 | 599 | |||
NM_001205098.1 | NP_001192027.1 | 8 | 1139 | |||
NM_026243.4 | NP_080519.2 | 4 | 621 | |||
GenBank mRNA | AK018574.1 | - | 4 | 620 | ||
AK018672.1 | - | 4 | 624 | |||
AK032009.1 | - | 6 | 600 | |||
AK033482.1 | - | 4 | 622 | |||
BC046987.1 | - | 4 | 609 | 6475851 |