Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026268.3 | NP_080544.1 | 2, 3 | 1291 | ||
GenBank mRNA | AK005062.1 | - | 2, 3 | 1292 | ||
AK009131.1 | - | 2, 3 | 1219 | |||
AK088468.1 | - | 2, 3 | 1292 | |||
AK088665.1 | - | 2, 3 | 1293 | |||
AK148192.1 | - | 2, 3 | 1292 | |||
AK151126.1 | - | 2, 3 | 1288 | |||
AK152773.1 | - | 2, 3 | 1288 | |||
AK159146.1 | - | 2, 3 | 1192 | |||
AK159502.1 | - | 2, 3 | 1292 | |||
AK159900.1 | - | 2, 3 | 1192 | |||
BC003869.1 | - | 2, 3 | 873 | 3491528 | ||
BC092272.1 | - | 2, 3 | 507 |