Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026310.3 | NP_080586.1 | 4 | 705 | ||
GenBank mRNA | AK003104.1 | - | 4 | 710 | ||
AK008680.1 | - | 4 | 706 | |||
AK089039.1 | - | 3 | 414 | |||
BC011425.1 | - | 4 | 680 | 3603233 |