Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026346.3 | NP_080622.1 | 9 | 1683 | ||
GenBank mRNA | AF441120.1 | - | 9 | 1729 | ||
AK014612.1 | - | 9 | 1636 | |||
AK014793.1 | - | 9 | 1665 | |||
BC027211.1 | - | 9 | 1617 | 3981597 |