Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930578I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026359.3 | NP_080635.2 | 4 | 752 | ||
GenBank mRNA | AK005778.1 | - | 4 | 749 | ||
AK016302.1 | - | 761 | ||||
BC048583.1 | - | 4 | 752 | 6706576 |