Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026369.2 | NP_080645.2 | 1, 2 | 336 | ||
GenBank mRNA | AK008097.1 | - | 1, 2 | 227 | ||
AK011348.1 | - | 1, 2 | 213 | |||
AK011911.1 | - | 1, 2 | 213 | |||
AK017669.1 | - | 1, 2 | 214 | |||
AK017985.1 | - | 1, 2 | 247 | |||
AK019149.1 | - | 1, 2 | 213 | |||
AK087427.1 | - | 1, 2 | 203 | |||
AK149756.1 | - | 1, 2 | 205 | |||
AK150443.1 | - | 1, 2 | 213 | |||
AK150703.1 | - | 1, 2 | 213 | |||
AK151490.1 | - | 1, 2 | 198 | |||
AK151654.1 | - | 1, 2 | 204 | |||
AK151656.1 | - | 1, 2 | 226 | |||
AK152281.1 | - | 1, 2 | 226 | |||
AK153262.1 | - | 1, 2 | 207 | |||
AK159407.1 | - | 1, 2 | 260 | |||
AK159438.1 | - | 1, 2 | 200 | |||
AK159738.1 | - | 1, 2 | 223 | |||
AK166284.1 | - | 1, 2 | 228 | |||
AK166335.1 | - | 1, 2 | 243 | |||
AK166725.1 | - | 1, 2 | 227 | |||
AK166919.1 | - | 1, 2 | 229 | |||
AK167916.1 | - | 1, 2 | 197 | |||
AK168264.1 | - | 1, 2 | 204 | |||
AK168986.1 | - | 1, 2 | 175 | |||
AK181325.1 | - | 1, 2 | 49 | |||
AK189385.1 | - | 1, 2 | 49 | |||
AK191774.1 | - | 1, 2 | 55 | |||
AK208127.1 | - | 1, 2 | 49 | |||
AK209531.1 | - | 1, 2 | 49 | |||
BC060143.1 | - | 1, 2 | 242 | 6411792 | ||
CF170085.1 | - | 1, 2 | 336 |