Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk2ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026373.4 | NP_080649.1 | 3 | 332 | ||
GenBank mRNA | AK003686.2 | - | 3 | 298 | ||
AK012121.1 | - | 3 | 335 | |||
AK018035.1 | - | 3 | 332 | |||
AK183323.1 | - | 1 | 87 | |||
BC025656.1 | - | 3 | 298 | 5342719 |