Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026380.3 | NP_080656.2 | 7 | 5029 | ||
GenBank mRNA | AK018215.1 | - | 1 | 822 | ||
BC056441.1 | - | 1 | 2035 | |||
BC057559.1 | - | 1 | 1461 | |||
BC065056.1 | - | 7 | 4962 | 5705033 | ||
BC094577.1 | - | 6 | 4645 | 6852696 |