Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026405.3 | NP_080681.1 | 2 | 376 | ||
GenBank mRNA | AB232627.1 | - | 2 | 257 | ||
AK012701.1 | - | 2 | 388 | |||
AK078874.1 | - | 2 | 68 | |||
AK088661.1 | - | 2 | 377 | |||
AK151543.1 | - | 2 | 350 | |||
AK159102.1 | - | 2 | 403 | |||
AK159901.1 | - | 2 | 403 | |||
AK163620.1 | - | 2 | 327 | |||
AK216759.1 | - | 1 | 16 | |||
AY135650.1 | - | 2 | 281 | |||
BC016409.1 | - | 294 | 4456902 | |||
BC055945.1 | - | 302 | 4161169 |