Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Knstrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026412.3 | NP_080688.2 | 4 | 564 | ||
GenBank mRNA | AK006328.1 | - | 4 | 565 | ||
AK012148.1 | - | 4 | 364 | |||
AK013184.1 | - | 4 | 312 | |||
AK028590.1 | - | 4 | 365 | |||
AK036374.1 | - | 4 | 353 | |||
BC031709.1 | - | 3 | 272 | 4021550 |