Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026443.4 | NP_080719.2 | 4 | 789 | ||
GenBank mRNA | AK006148.1 | - | 4 | 710 | ||
AK010971.1 | - | 4 | 746 | |||
AK012058.1 | - | 4 | 756 | |||
AK078381.1 | - | 4 | 790 | |||
BC012242.1 | - | 1 | 203 | |||
BC019205.1 | - | 4 | 799 | |||
BC058737.1 | - | 4 | 739 | 5400435 |