Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026444.3 | NP_080720.1 | 5 | 415 | ||
GenBank mRNA | AB056479.1 | - | 5 | 329 | ||
AK010298.1 | - | 5 | 504 | |||
AK012151.1 | - | 5 | 446 | |||
AK145643.1 | - | 5 | 447 | |||
AK149990.1 | - | 5 | 514 | |||
AK163273.1 | - | 5 | 440 | |||
AK166814.1 | - | 5 | 450 | |||
AK167125.1 | - | 5 | 447 | |||
BC013554.1 | - | 5 | 430 | 4237940 | ||
BC029754.1 | - | 5 | 415 | 2655044 |