Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161800.1 | NP_001155272.1 | 1, 2 | 286 | ||
NM_026448.3 | NP_080724.2 | 1, 2 | 286 | |||
GenBank mRNA | AK012326.1 | - | 1, 2 | 267 | ||
AK082520.1 | - | 1, 2 | 229 | |||
AK083358.1 | - | 1, 2 | 272 | |||
AK089072.1 | - | 1, 2 | 271 | |||
BB841624.1 | - | 1, 2 | 287 | |||
BC029154.1 | - | 1, 2 | 253 | 4952319 |