Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026470.3 | NP_080746.3 | 5 | 580 | ||
GenBank mRNA | AK051574.1 | - | 5 | 581 | ||
AK053170.1 | - | 5 | 554 | |||
AK132771.1 | - | 5 | 554 | |||
AK153098.1 | - | 5 | 457 |