Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026369.2 | NP_080645.2 | 3 | 528 | ||
GenBank mRNA | AK008097.1 | - | 3 | 419 | ||
AK011348.1 | - | 3 | 405 | |||
AK011911.1 | - | 3 | 405 | |||
AK017669.1 | - | 3 | 406 | |||
AK017985.1 | - | 3 | 438 | |||
AK019149.1 | - | 3 | 405 | |||
AK087427.1 | - | 3 | 395 | |||
AK149756.1 | - | 3 | 397 | |||
AK150443.1 | - | 3 | 405 | |||
AK150703.1 | - | 3 | 405 | |||
AK151654.1 | - | 3 | 396 | |||
AK151656.1 | - | 3 | 418 | |||
AK152281.1 | - | 3 | 418 | |||
AK153262.1 | - | 3 | 399 | |||
AK159407.1 | - | 3 | 452 | |||
AK159438.1 | - | 3 | 392 | |||
AK159738.1 | - | 3 | 415 | |||
AK166284.1 | - | 3 | 420 | |||
AK166335.1 | - | 3 | 435 | |||
AK166725.1 | - | 3 | 419 | |||
AK166919.1 | - | 3 | 421 | |||
AK167144.1 | - | 2 | 175 | |||
AK167916.1 | - | 3 | 389 | |||
AK168264.1 | - | 3 | 396 | |||
AK168986.1 | - | 3 | 367 | |||
AK189385.1 | - | 3 | 244 | |||
AK191774.1 | - | 3 | 247 | |||
AK208221.1 | - | 2 | 195 | |||
AK209531.1 | - | 3 | 241 | |||
BC060143.1 | - | 3 | 434 | 6411792 | ||
CF170085.1 | - | 3 | 528 |