Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026380.3 | NP_080656.2 | 7 | 5089 | ||
GenBank mRNA | AK018215.1 | - | 1 | 882 | ||
AK018337.1 | - | 1 | 1065 | |||
BC056441.1 | - | 1 | 2095 | |||
BC057559.1 | - | 1 | 1521 | |||
BC065056.1 | - | 7 | 5022 | 5705033 | ||
BC094577.1 | - | 6 | 4705 | 6852696 |