Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026382.2 | NP_080658.2 | 2 | 284 | ||
GenBank mRNA | AK007431.1 | - | 2 | 273 | ||
AK012916.1 | - | 2 | 279 | |||
AK018315.1 | - | 2 | 279 | |||
AK036713.1 | - | 2 | 284 | |||
AK087789.1 | - | 2 | 279 | |||
AK172155.1 | - | 2 | 273 | |||
BC021443.1 | - | 2 | 267 | 5038605 | ||
BC037073.1 | - | 2 | 274 | |||
BY263553.1 | - | 2 | 287 |