Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enoph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163035.1 | NP_001156507.1 | 3 | 425 | ||
NM_026421.3 | NP_080697.2 | 3 | 425 | |||
GenBank mRNA | AK009954.1 | - | 3 | 408 | ||
AK079062.1 | - | 3 | 419 | |||
AK080714.1 | - | 3 | 376 | |||
AK082747.1 | - | 3 | 400 | |||
AK152021.1 | - | 3 | 400 | |||
AK159356.1 | - | 3 | 398 | |||
AK167575.1 | - | 3 | 436 | |||
AK177339.1 | - | 2 | 36 | |||
AK178746.1 | - | 2 | 36 | |||
BC021429.1 | - | 3 | 352 | 5030617 | ||
CX235072.1 | - | 3 | 343 |