Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026423.4 | NP_080699.3 | 2 | 477 | ||
GenBank mRNA | AK007709.1 | - | 2 | 438 | ||
AK010538.1 | - | 2 | 449 | |||
AK013882.1 | - | 2 | 446 | |||
AK047287.1 | - | 2 | 480 | |||
AK144807.1 | - | 2 | 418 | |||
AK164635.1 | - | 2 | 477 | |||
AK164894.1 | - | 2 | 446 | |||
BC025049.1 | - | 2 | 444 | 4951045 |