Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026441.4 | NP_080717.2 | 5 | 871 | ||
GenBank mRNA | AK007394.1 | - | 5 | 813 | ||
AK013238.1 | - | 5 | 812 | |||
AK046247.1 | - | 5 | 813 | |||
AK075978.1 | - | 5 | 876 | |||
BC019191.1 | - | 5 | 819 | 5029503 |