Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026442.3 | NP_080718.1 | 3 | 298 | ||
GenBank mRNA | AK002667.1 | - | 3 | 195 | ||
AK008567.1 | - | 3 | 217 | |||
AK011760.1 | - | 3 | 199 | |||
AK020626.1 | - | 3 | 182 | |||
AK168728.1 | - | 3 | 245 | |||
BC048547.1 | - | 3 | 279 | |||
BC056653.1 | - | 3 | 172 | 5068125 | ||
CJ170455.1 | - | 3 | 299 |