Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026447.4 | NP_080723.3 | 3, 4 | 645 | ||
NM_198931.3 | NP_945149.2 | 3, 4 | 649 | |||
GenBank mRNA | AB474373.1 | - | 3, 4 | 684 | ||
AK013149.1 | - | 3, 4 | 578 | |||
AK046387.1 | - | 544 | ||||
AK087999.1 | - | 3, 4 | 683 | |||
AY332616.1 | - | 3, 4 | 652 | |||
BC058248.1 | - | 3, 4 | 571 |