Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1600012H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083880.1 | NP_001077349.1 | 2 | 470 | ||
NM_001083881.1 | NP_001077350.1 | 2 | 540 | |||
NM_001083882.1 | NP_001077351.1 | 1 | 722 | |||
NM_026451.2 | NP_080727.2 | 2 | 475 | |||
GenBank mRNA | AK005427.1 | - | 2 | 452 | ||
AK017276.1 | - | 1 | 652 | |||
AK018298.1 | - | 2 | 398 | |||
AK028052.1 | - | 2 | 476 | |||
AK076083.1 | - | 2 | 432 | |||
AK076441.1 | - | 679 | ||||
AK076476.1 | - | 2 | 395 | |||
AK083064.1 | - | 1 | 663 | |||
AK150772.1 | - | 2 | 495 | |||
AK167490.1 | - | 2 | 482 | |||
BB610149.1 | - | 2 | 476 | |||
BC027313.1 | - | 2 | 454 | 3710655 | ||
BC043303.1 | - | 1 | 683 | |||
BC043449.1 | - | 1 | 683 | |||
BC054359.1 | - | 2 | 472 | |||
BC071250.1 | - | 2 | 387 |