Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026348.3 | NP_080624.1 | 9 | 1466 | ||
GenBank mRNA | AK015557.1 | - | 9 | 1491 | ||
AK045037.1 | - | 9 | 1467 | |||
AK165072.1 | - | 7 | 1322 |