Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026358.3 | NP_080634.2 | 4 | 1058 | ||
GenBank mRNA | AF228502.1 | - | 4 | 1037 | ||
AK006339.1 | - | 4 | 1020 | |||
AK016342.1 | - | 4 | 1031 | |||
AK151401.1 | - | 4 | 1052 | |||
BC012511.1 | - | 4 | 1051 | 4500810 | ||
BC022740.1 | - | 4 | 1212 | 4511440 | ||
BC049556.1 | - | 4 | 1020 | 6772062 |