Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plgrkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026362.2 | NP_080638.1 | 6 | 533 | ||
GenBank mRNA | AK003543.1 | - | 4 | 294 | ||
AK017215.1 | - | 6 | 534 | |||
AK168454.1 | - | 6 | 509 | |||
AK170700.1 | - | 7 | 607 | |||
BC024953.1 | - | 6 | 492 | 4951101 |