Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026370.1 | NP_080646.1 | 6 | 737 | ||
GenBank mRNA | AK003264.1 | - | 6 | 737 | ||
AK008418.1 | - | 6 | 724 | |||
AK018002.1 | - | 6 | 733 | |||
AK132000.1 | - | 6 | 738 | |||
AK146710.1 | - | 6 | 739 | |||
BC036284.1 | - | 6 | 745 | 5136304 |