Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Loh12cr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170479.1 | NP_001163950.1 | 4 | 1195 | ||
NM_026371.3 | NP_080647.2 | 4 | 1337 | |||
GenBank mRNA | AK007423.1 | - | 4 | 1194 | ||
AK018019.1 | - | 4 | 1253 | |||
AK151342.1 | - | 4 | 1203 | |||
AK169615.1 | - | 4 | 1209 | |||
BC023059.1 | - | 4 | 1296 | 5355560 |