Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000229.1 | NP_000220.1 | 1 | 136 | ||
GenBank mRNA | BC014781.1 | - | 1 | 126 | 4155094 | |
BT009748.1 | - | 1 | 125 | |||
M12625.1 | - | 1 | 392 | |||
M26268.1 | - | 1 | 78 | |||
X06537.1 | - | 1 | 228 |
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145961.1 | NP_001139433.1 | 24 | 4244 | ||
NM_001145962.1 | NP_001139434.1 | 23 | 4167 | |||
NM_001145963.1 | NP_001139435.1 | 24 | 4141 | |||
NM_001145964.1 | NP_001139436.1 | 24 | 4132 | |||
NM_005072.4 | NP_005063.1 | 24 | 4262 | |||
GenBank mRNA | AK310027.1 | - | 1 | 544 |