Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026405.3 | NP_080681.1 | 3 | 1233 | ||
GenBank mRNA | AK012701.1 | - | 3 | 1245 | ||
AK078874.1 | - | 3 | 925 | |||
AK088661.1 | - | 3 | 1234 | |||
AK151543.1 | - | 3 | 1207 | |||
AK159102.1 | - | 3 | 1260 | |||
AK159901.1 | - | 3 | 1260 | |||
AK163620.1 | - | 3 | 1184 | |||
AY135650.1 | - | 3 | 1136 | |||
BC016409.1 | - | 1151 | 4456902 | |||
BC055945.1 | - | 1159 | 4161169 |