Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026418.2 | NP_080694.1 | 4 | 339 | ||
GenBank mRNA | AK009283.1 | - | 4 | 340 | ||
AK009686.1 | - | 4 | 341 | |||
AK017314.1 | - | 4 | 340 | |||
AK021202.1 | - | 4 | 340 | |||
AK088009.1 | - | 4 | 338 | |||
BC109178.1 | - | 4 | 324 | 40049527 | ||
BC109179.1 | - | 4 | 331 |