Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026444.3 | NP_080720.1 | 11 | 1607 | ||
GenBank mRNA | AK012104.1 | - | 5 | 785 | ||
AK012151.1 | - | 11 | 1638 | |||
AK149990.1 | - | 11 | 1706 | |||
AK163273.1 | - | 11 | 1632 | |||
BC013554.1 | - | 11 | 1622 | 4237940 | ||
BC029754.1 | - | 11 | 1607 | 2655044 |