Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1600012H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083880.1 | NP_001077349.1 | 2 | 1022 | ||
NM_001083881.1 | NP_001077350.1 | 2 | 1092 | |||
NM_001083882.1 | NP_001077351.1 | 1 | 1274 | |||
NM_026451.2 | NP_080727.2 | 2 | 1027 | |||
GenBank mRNA | AK005427.1 | - | 2 | 1004 | ||
AK017276.1 | - | 1 | 1204 | |||
AK018298.1 | - | 2 | 948 | |||
AK028052.1 | - | 2 | 1030 | |||
AK076083.1 | - | 2 | 984 | |||
AK076441.1 | - | 1231 | ||||
AK076476.1 | - | 2 | 947 | |||
AK083064.1 | - | 1 | 1215 | |||
AK150772.1 | - | 2 | 1047 | |||
AK167490.1 | - | 2 | 1034 | |||
BC027313.1 | - | 2 | 1006 | 3710655 | ||
BC043303.1 | - | 1 | 1235 | |||
BC043449.1 | - | 1 | 1235 | |||
BC054359.1 | - | 2 | 1024 | |||
BC071250.1 | - | 2 | 939 |