Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026473.2 | NP_080749.2 | 4 | 1085 | ||
GenBank mRNA | AK009958.1 | - | 4 | 1093 | ||
AK015255.1 | - | 1 | 351 | |||
AK144815.1 | - | 4 | 1100 | |||
AK151643.1 | - | 4 | 1082 | |||
BC008225.1 | - | 4 | 1085 | 2631779 |