Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccny | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026484.3 | NP_080760.2 | 4 | 761 | ||
GenBank mRNA | AK017493.1 | - | 4 | 593 | ||
AK028696.1 | - | 4 | 762 | |||
AK076556.1 | - | 4 | 291 | |||
AK215714.1 | - | 2 | 55 | |||
BC023321.1 | - | 5 | 679 | 3995040 | ||
BC035524.1 | - | 3 | 516 | 4974858 |