Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026494.3 | NP_080770.2 | 3 | 937 | ||
GenBank mRNA | AK006290.1 | - | 3 | 449 | ||
AK018256.1 | - | 3 | 909 | |||
AK159952.1 | - | 3 | 937 | |||
AK213637.1 | - | 1 | 164 | |||
BC021894.1 | - | 3 | 874 | 3494382 |