Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037098.1 | NP_001032175.1 | 2 | 1518 | ||
NM_026495.3 | NP_080771.3 | 2 | 995 | |||
GenBank mRNA | AK002651.1 | - | 2 | 969 | ||
AK043047.1 | - | 2 | 943 | |||
AK147293.1 | - | 2 | 995 | |||
AK147310.1 | - | 2 | 1099 | |||
AK157946.1 | - | 2 | 1102 | |||
AK184562.1 | - | 1 | 83 | |||
BC022103.1 | - | 2 | 977 | 5067063 | ||
BC070453.1 | - | 2 | 1298 |