Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000243.2 | NP_000234.1 | 1, 2 | 311 | ||
XM_017023236.1 | XP_016878725.1 | 1, 2 | 452 | |||
GenBank mRNA | AF018080.1 | - | 1, 2 | 312 | ||
AK298660.1 | - | 1, 2 | 305 | |||
BC101511.1 | - | 1, 2 | 296 | 8069017 | ||
BC101537.1 | - | 1, 2 | 296 | 8069043 | ||
FJ785721.1 | - | 1, 2 | 271 | |||
Y14441.1 | - | 1, 2 | 300 |