Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700060E02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026528.3 | NP_080804.1 | 2 | 238 | ||
GenBank mRNA | AK003732.1 | - | 2 | 234 | ||
AK012451.1 | - | 2 | 202 | |||
AK012460.1 | - | 2 | 233 | |||
AK012486.1 | - | 2 | 239 | |||
AK148686.1 | - | 2 | 180 | |||
AK160555.1 | - | 2 | 202 | |||
AK160696.1 | - | 2 | 240 | |||
AK162070.1 | - | 2 | 190 | |||
AK162433.1 | - | 2 | 167 | |||
BC023932.1 | - | 2 | 217 | |||
BC048357.1 | - | 2 | 233 | 3979120 | ||
BC096571.1 | - | 2 | 221 | 963861 |