Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026542.3 | NP_080818.1 | 5 | 365 | ||
GenBank mRNA | AK002381.1 | - | 5 | 323 | ||
AK007276.1 | - | 5 | 344 | |||
AK007934.1 | - | 5 | 319 | |||
AK010272.1 | - | 5 | 344 | |||
AK019396.1 | - | 5 | 357 | |||
AK078417.1 | - | 5 | 357 | |||
AK084116.1 | - | 5 | 356 | |||
AK160922.1 | - | 5 | 310 | |||
AK161289.1 | - | 5 | 357 | |||
BC115661.1 | - | 5 | 265 | 40103267 | ||
BC115662.1 | - | 5 | 272 | 40103268 | ||
BY291748.1 | - | 5 | 366 |