Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026559.3 | NP_080835.1 | 1 | 183 | ||
GenBank mRNA | AK004219.1 | - | 1 | 191 | ||
AK007595.1 | - | 1 | 187 | |||
AK013103.1 | - | 1 | 126 | |||
AK076425.1 | - | 1 | 183 | |||
AK169882.1 | - | 1 | 126 | |||
BC030344.1 | - | 1 | 181 | 3673521 |