Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026562.2 | NP_080838.1 | 2 | 342 | ||
GenBank mRNA | AK006754.1 | - | 2 | 128 | ||
AK007245.1 | - | 2 | 343 | |||
AK049116.1 | - | 2 | 450 | |||
BC006866.1 | - | 2 | 401 | 3595113 |