Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026483.2 | NP_080759.2 | 6, 7 | 1361 | ||
GenBank mRNA | AK008785.1 | - | 3, 4 | 291 | ||
AK013337.1 | - | 6, 7 | 1349 | |||
BC018177.1 | - | 6, 7 | 1355 | |||
BC049270.1 | - | 6, 7 | 1337 | 3988645 |