Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026485.2 | NP_080761.2 | 6 | 580 | ||
GenBank mRNA | AK005300.1 | - | 6 | 550 | ||
AK017745.1 | - | 6 | 574 | |||
AK131116.1 | - | 1 | 2219 | |||
AK146027.1 | - | 6 | 596 | |||
AK185880.1 | - | 2 | 140 | |||
BC005574.1 | - | 6 | 606 | 3967961 | ||
BU510197.1 | - | 6 | 660 | |||
CX200611.1 | - | 6 | 565 |