Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026498.2 | NP_080774.2 | 3 | 456 | ||
GenBank mRNA | AB049945.1 | - | 3 | 180 | ||
AK002993.1 | - | 3 | 293 | |||
AK152027.1 | - | 3 | 247 | |||
AK160207.1 | - | 3 | 236 | |||
BC005701.1 | - | 3 | 198 | |||
BC028279.1 | - | 3 | 248 | 5370270 |