Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwilch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026507.4 | NP_080783.3 | 3 | 205 | ||
GenBank mRNA | AK009559.1 | - | 3 | 176 | ||
AK012004.1 | - | 4 | 248 | |||
AK019825.1 | - | 3 | 183 | |||
AK050917.1 | - | 3 | 170 | |||
AK186156.1 | - | 57 | ||||
BB839968.1 | - | 3 | 206 | |||
BC027435.1 | - | 3 | 153 | 2812472 |