Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170486.1 | NP_001163957.1 | 2 | 146 | ||
NM_026552.3 | NP_080828.1 | 3 | 265 | |||
GenBank mRNA | AJ278129.1 | - | 3 | 191 | ||
AK017993.1 | - | 3 | 266 | |||
AK018069.1 | - | 3 | 203 | |||
AK030840.1 | - | 1 | 3848 | |||
AK039215.1 | - | 3 | 213 | |||
AK076248.1 | - | 3 | 213 | |||
AK077325.1 | - | 3 | 187 | |||
AK133626.1 | - | 3 | 213 | |||
AK136082.1 | - | 3 | 208 | |||
AK150690.1 | - | 3 | 199 | |||
AK153380.1 | - | 3 | 197 | |||
AK159410.1 | - | 2 | 94 | |||
AK166440.1 | - | 3 | 187 | |||
AK193553.1 | - | 2 | 136 | |||
AK194624.1 | - | 2 | 136 | |||
AK194784.1 | - | 2 | 136 | |||
AK197517.1 | - | 2 | 136 | |||
BC015280.1 | - | 2 | 177 | |||
BC055309.1 | - | 2 | 176 | 6529255 |